Please login to your online account to display your discounted pricing

Alfa Aesar™ 12-Aminododecanoic acid, 96%

$532.59 - $532.59

Chemical Identifiers

Chemical Name or Material 12-Aminododecanoic acid
CAS 693-57-2
Molecular Formula H2N(CH2)11CO2H
Formula Weight 215.34
MDL Number MFCD00008153
Synonym 12-Aminolauric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2512114

 
alfa aesar™
B2512114
25g Each for $532.59
Description & Specifications

Specifications

Formula Weight 215.34
Chemical Name or Material 12-Aminododecanoic acid
TSCA TSCA
Melting Point 185° to 187°C
Assay 96%
Quantity 25g
Molecular Formula H2N(CH2)11CO2H
MDL Number MFCD00008153
CAS 693-57-2
Synonym 12-Aminolauric acid