Please login to your online account to display your discounted pricing
Thermo Scientific™ 11mm Non-Assembled Clear Crimp Top Autosampler Vial Kits
Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear glass vials and 100 crimp top caps with pre-assembled septa.
Manufacturer: thermo scientific™  SPL*
 View more versions of this product
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Catalog No. 03-341-938
  • / Pack of 100 Each
Description & Specifications
Specifications
Material Glass
Color Clear
Patched Yes
No. per Pack 100
Item Description IS Kit, Wide Opening Crimp Top Vial for Spark LC Autosampler
Save time during sample preparation and reduce the risk of contamination using Thermo Scientific™ 11mm Crimp Top Clear Vial Convenience kits (Unassembled). Each kit includes 100 clear glass vials and 100 crimp top caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures while keeping matching supplies together. Clear trays to make it easy to keep track of available supplies without opening containers. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.