Please login to your online account to display your discounted pricing
Thermo Scientific™ 11mm Assembled Amber Autosampler Vial Kits
Save time and reduce the risk of contamination using this convenience kit. It includes 100 amber vials pre-assembled with 100 screw thread caps and septa.
$78.59 - $78.59
Specifications
Closure Color Silver
Closure Material Aluminum
Description Amber Glass I-D; Silver seal, PTFE/Red rubber septa
Closure Type Crimped
Color Amber
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Vial Closure Size Vial Volume (Metric) Price Quantity    
03-376-451
thermo scientific™
C40112WCV
11mm 2mL Pack of 100 Each for $78.59
Description & Specifications
Specifications
Closure Color Silver
Closure Material Aluminum
Description Amber Glass I-D; Silver seal, PTFE/Red rubber septa
Closure Type Crimped
Color Amber
Material Glass
No. per Pack 100
Patched Yes
Type Assembled Vial Kit
Item Description Assembled Kit, Wide Opening Crimp Top Vial, Nitrogen purged
Septa PTFE/Red rubber
Vial Closure Size 11mm
Vial Volume (Metric) 2mL
Save time during sample preparation and reduce the risk of contamination. Each Thermo Scientific™ 11mm Amber Vial Convenience kits (Assembled) includes 100 amber glass vials pre-assembled with 100 screw thread caps and septa. They are packaged in convenient vial trays with clear covers or in economical polybags. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.