Please login to your online account to display your discounted pricing

Alfa Aesar™ 11H-Perfluoroundecanoic acid, 95%


Quantity 100g

Manufacturer: alfa aesar™  L1675922

 View more versions of this product

Catalog No. AAL1675922

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 546.11
Chemical Name or Material 11H-Perfluoroundecanoic acid
Assay 95%
MDL Number MFCD00042431
CAS 1765-48-6
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA TSCA
Molecular Formula H(CF2)10CO2H
UN Number UN3261
Synonym 11H-Eicosafluoroundecanoic acid
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >100°C (212°F)
Melting Point 102° to 104°C
Quantity 100g

Specifications

Chemical Name or Material 11H-Perfluoroundecanoic acid
CAS 1765-48-6
Molecular Formula H(CF2)10CO2H
Formula Weight 546.11
MDL Number MFCD00042431
UN Number UN3261
Synonym 11H-Eicosafluoroundecanoic acid
Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

SDS & Safety Documents