Please login to your online account to display your discounted pricing

11-Chloro-1-undecene 97%, ACROS Organics™

$254.44 - $254.44

Chemical Identifiers

Chemical Name or Material 11-Chloro-1-undecene
CAS 872-17-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H21Cl
Linear Formula CH2CH(CH2)9Cl
Formula Weight 188.74
MDL Number MFCD00671354
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380840250

  SDS
fisher scientific acros
AC380840250
Glass bottle 25mL Each for $254.44
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 188.74
Physical Form Liquid
Quantity 25mL
Color Clear colorless
Density 0.9950g/mL
CAS Min % 96.0
CAS Max % 100.0
Linear Formula CH2CH(CH2)9Cl
Molecular Formula C11H21Cl
MDL Number MFCD00671354
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Chemical Name or Material 11-Chloro-1-undecene
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4500 to 1.4520 (20°C, 589nm)
CAS 872-17-3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.