missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 11-Chloro-1-undecene, 97%

Manufacturer:  Alfa Aesar™ H53449AC

 View more versions of this product

Catalog No. AAH53449AC



Description

Description

Applications
11-Chloro-1-undecene is used in copolymerizations of propylene , preparation of unsaturated fatty acids, terpolymerizations with ethylene and propylene catalyzed by 2/mao.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

11-Chloro-1-undecene
108°C to 109°C (6mmHg)
872-17-3
Cl(CH2)9CH=CH2
1-undecene, 11-chloro
C=CCCCCCCCCCCl
188.739
188.74
104°C (219°F)
25mL
C11H21Cl
MFCD00671354
PSEVKFKRYVAODC-UHFFFAOYSA-N
11-chloroundec-1-ene
543805
97%

Chemical Identifiers

872-17-3
188.739
PSEVKFKRYVAODC-UHFFFAOYSA-N
543805
C=CCCCCCCCCCCl
C11H21Cl
MFCD00671354
1-undecene, 11-chloro
11-chloroundec-1-ene
Documents