missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 11-Bromoundecanol, 97%

Manufacturer:  Alfa Aesar™ L1444814

 View more versions of this product

Catalog No. AAL1444814



Description

Description

Applications
11-Bromoundecanol was used in the preparation of 11-mercapto-1-undecanol1 and 1-(2-acryloyloxyundecyl)-3-methylimidazolium bromide. It was also used in one-pot synthesis of graft copolymers based on a metathesis skeleton with poly(methyl methacrylate) grafts.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

11-Bromoundecanol
165°C to 170°C (1mmHg)
25g
C11H23BrO
1741180
XFGANBYCJWQYBI-UHFFFAOYSA-N
11-bromoundecan-1-ol
74163
97%
>110°C (230°F)
46°C to 49°C
1611-56-9
MFCD00004752
11-bromo-1-undecanol, 11-bromoundecanol, 1-undecanol, 11-bromo, 1-bromo-11-hydroxyundecane, undecamethylene bromohydrin, 11-bromoundecyl alcohol, 11-bromo-l-undecanol, 11-bromoundecane-1-ol, 11-bromanylundecan-1-ol, acmc-1br3a
C(CCCCCO)CCCCCBr
251.208
251.21

Chemical Identifiers

1611-56-9
251.208
XFGANBYCJWQYBI-UHFFFAOYSA-N
74163
C(CCCCCO)CCCCCBr
C11H23BrO
MFCD00004752
11-bromo-1-undecanol, 11-bromoundecanol, 1-undecanol, 11-bromo, 1-bromo-11-hydroxyundecane, undecamethylene bromohydrin, 11-bromoundecyl alcohol, 11-bromo-l-undecanol, 11-bromoundecane-1-ol, 11-bromanylundecan-1-ol, acmc-1br3a
11-bromoundecan-1-ol
Documents