Please login to your online account to display your discounted pricing

11-Bromoundecanoic acid, 93%, Tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material 11-Bromoundecanoic acid
Name Note tech., 93%
CAS 341259
CAS Min % 91.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥91%
Assay Percent Range 93%
Molecular Formula C11H21BrO2
Linear Formula Br(CH2)10CO2H
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC173222500

  SDS
thermo fisher scientific
173222500
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥91%
Formula Weight 265.19
Purity Grade Technical Grade
Physical Form Crystalline Solid
Quantity 250g
Color Beige to light brown
TSCA TSCA
EINECS Number 220-602-9
Melting Point 46° to 51°C
Beilstein 02, II, 315
CAS Max % 100.0
CAS 341259
CAS Min % 91.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C11H21BrO2
Name Note tech., 93%
Linear Formula Br(CH2)10CO2H
Chemical Name or Material 11-Bromoundecanoic acid
Assay Percent Range 93%
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.