Learn More
11-Bromo-1-undecanol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 293440500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
11-Bromo-1-undecanol | |
1611-56-9 | |
100.0 | |
White to Beige | |
>110°C | |
97% | |
C11H23BrO | |
MFCD00004752 | |
01, IV, 1837 | |
XFGANBYCJWQYBI-UHFFFAOYSA-N | |
11-bromoundecan-1-ol | |
74163 | |
97% |
97% | |
96.0 | |
42°C to 47°C | |
165°C to 170°C (1.0 mmHg) | |
Authentic | |
Glass bottle | |
Br(CH2)11OH | |
50 g | |
11-bromo-1-undecanol, 11-bromoundecanol, 1-undecanol, 11-bromo, 1-bromo-11-hydroxyundecane, undecamethylene bromohydrin, 11-bromoundecyl alcohol, 11-bromo-l-undecanol, 11-bromoundecane-1-ol, 11-bromanylundecan-1-ol, acmc-1br3a | |
OCCCCCCCCCCCBr | |
251.21 | |
251.21 | |
Crystalline Powder or Crystals |
Chemical Identifiers
1611-56-9 | |
251.21 | |
XFGANBYCJWQYBI-UHFFFAOYSA-N | |
74163 | |
OCCCCCCCCCCCBr |
C11H23BrO | |
MFCD00004752 | |
11-bromo-1-undecanol, 11-bromoundecanol, 1-undecanol, 11-bromo, 1-bromo-11-hydroxyundecane, undecamethylene bromohydrin, 11-bromoundecyl alcohol, 11-bromo-l-undecanol, 11-bromoundecane-1-ol, 11-bromanylundecan-1-ol, acmc-1br3a | |
11-bromoundecan-1-ol |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 216-554-3
TSCA : TSCA
RUO – Research Use Only