Please login to your online account to display your discounted pricing

10X PBS Liquid Concentrate, Sterile, OmniPur™, Calbiochem™, EMD Millipore

Each liter makes 10L of 1X PBS Buffer

$103.94 - $103.94

Chemical Identifiers

Analyte or Component Names Water, Sodium Chloride, Sodium Phosphate (Dibasic, Anhydrous), Potassium Chloride, Potassium Phosphate (Monobasic)
CAS 7732-18-5
Chemical Name or Material 10X PBS Liquid Concentrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

65061L

 
emd millipore corp
65061L
Each for $103.94
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material 10X PBS Liquid Concentrate
Analyte or Component Names Water, Sodium Chloride, Sodium Phosphate (Dibasic, Anhydrous), Potassium Chloride, Potassium Phosphate (Monobasic)
CAS 7732-18-5
CAS 7558-79-4
RNase None Detected
CAS 7447-40-7
CAS 7778-77-0
DNase None Detected
Synonym Phosphate Buffered Saline
CAS 7647-14-5