Fisherbrand™ Screw Vial Convenience Kits with Caps
Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear or amber glass vials and 100 screw thread caps with septa.
Manufacturer:  Fisherbrand™ 13622199
 View more versions of this product
Catalog No. 13-622-199
Description
Description
Fisherbrand™ Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
  • Convenience Kits with 2mL, 12 × 32mm vials
  • I-D vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
  • Blue caps with your choice of vial and preassembled seals with septa
  • Packaged in two-compartment benchtop storage tray
  • All items remain dust-free
  • Caps with pre-inserted septa are not attached to vials (packed in separate polybag)
Specifications
Specifications
Light Blue
Screw
Chromatography
Yes
Red PTFE/White Silicone
Clear
10mm
Clear Glass w/I-D Patch, Light Blue DP Cap w/ Red PTFE/White Silicone Septum
Vials and Closures
100
Autosampler Vial
Glass, PTFE, Silicone
Documents