Learn More
10-Undecen-1-ol, 98+%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 388990250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
10-Undecen-1-ol | |
97.5 | |
-3.0°C | |
0.8460g/mL | |
93°C | |
98% min. (GC) | |
C11H22O | |
H2C=CH(CH2)9OH | |
25 g | |
0.846 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol | |
C=CCCCCCCCCCO | |
170.29 | |
170.29 | |
Liquid |
112-43-6 | |
100.0 | |
Colorless | |
245.0°C to 248.0°C | |
Authentic | |
Glass bottle | |
1.4460 to 1.4530 | |
MFCD00004750 | |
01,452 | |
10-undecen-1-ol, undecylenic alcohol, undecylenyl alcohol, 10-undecenol, 1-undecen-11-ol, 10-undecylen-1-ol, 11-hydroxy-1-undecene, omega-undecenyl alcohol, undecen-1-ol, .omega.-undecenyl alcohol | |
GIEMHYCMBGELGY-UHFFFAOYSA-N | |
undec-10-en-1-ol | |
8185 | |
98+% |
Chemical Identifiers
112-43-6 | |
170.29 | |
GIEMHYCMBGELGY-UHFFFAOYSA-N | |
8185 | |
C=CCCCCCCCCCO |
C11H22O | |
MFCD00004750 | |
10-undecen-1-ol, undecylenic alcohol, undecylenyl alcohol, 10-undecenol, 1-undecen-11-ol, 10-undecylen-1-ol, 11-hydroxy-1-undecene, omega-undecenyl alcohol, undecen-1-ol, .omega.-undecenyl alcohol | |
undec-10-en-1-ol |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 203-971-
RUO – Research Use Only