Please login to your online account to display your discounted pricing

Alfa Aesar™ 10-Phenyl-1-decanol, 96%

$114.36 - $418.69

Chemical Identifiers

Chemical Name or Material 10-Phenyl-1-decanol
CAS 62607-69-6
Molecular Formula C6H5(CH2)10OH
Formula Weight 234.39
MDL Number MFCD00014426
Beilstein 1955067
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1026906

 
alfa aesar™
L1026906
5g Each for $418.69

AAL1026903

 
alfa aesar™
L1026903
1g Each for $114.36
Description & Specifications

Specifications

Formula Weight 234.39
MDL Number MFCD00014426
Assay 96%
Molecular Formula C6H5(CH2)10OH
CAS 62607-69-6
Beilstein 1955067
Boiling Point 145° to 147°C (0.5mmHg)
Melting Point 35° to 37°C
Quantity 5g
Chemical Name or Material 10-Phenyl-1-decanol