missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
10-Methylphenothiazine 98.0+%, TCI America™
$84.60 - $84.60
Chemical Identifiers
CAS | 1207-72-3 |
---|---|
Molecular Formula | C13H11NS |
Molecular Weight (g/mol) | 213.30 |
MDL Number | MFCD00041836 |
InChI Key | QXBUYALKJGBACG-UHFFFAOYSA-N |
Synonym | 10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci |
PubChem CID | 71015 |
IUPAC Name | 10-methyl-10H-phenothiazine |
SMILES | CN1C2=CC=CC=C2SC2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M25175G
|
TCI America
M25175G |
5 g |
Each for $84.60
|
|
|||||
Chemical Identifiers
1207-72-3 | |
213.30 | |
QXBUYALKJGBACG-UHFFFAOYSA-N | |
71015 | |
CN1C2=CC=CC=C2SC2=CC=CC=C12 |
C13H11NS | |
MFCD00041836 | |
10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci | |
10-methyl-10H-phenothiazine |
Specifications
1207-72-3 | |
White-Yellow | |
MFCD00041836 | |
10-methyl-10h-phenothiazine, n-methylphenothiazine, 10h-phenothiazine, 10-methyl, phenothiazine, 10-methyl, acmc-1bo1o, bidd:gt0308, phenothiazide methyl derivative, 10-methylphenothiazine, 10-methyl-10h-phenothiazine #, phenothiazine, 10-methyl-8ci | |
CN1C2=CC=CC=C2SC2=CC=CC=C12 | |
213.30 | |
213.30 | |
Crystalline Powder |
102°C | |
C13H11NS | |
5 g | |
QXBUYALKJGBACG-UHFFFAOYSA-N | |
10-methyl-10H-phenothiazine | |
71015 | |
≥98.0% (GC) | |
10-Methylphenothiazine |
Safety and Handling
TSCA : Yes