missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

10-Hydroxydecanoic acid, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6074303

 View more versions of this product

Catalog No. AAH6074303



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

10-Hydroxydecanoic acid
75°C to 77°C
HO(CH2)9CO2H
1 g
YJCJVMMDTBEITC-UHFFFAOYSA-N
10-hydroxydecanoic acid
74300
188.26
1679-53-4
C10H20O3
MFCD00010510
10-hydroxydecanoate, decanoic acid, 10-hydroxy, unii-np03xo416b, 10-hydroxy capric acid, 10-hydroxy-decanoic acid, 1-hydroxydecanoicacid, 10-oh-capric acid, 10-hydroxycapric acid, 10-oh-decanoic acid, 10-hydroxydecanoicacid
OCCCCCCCCCC(O)=O
188.27
CHEBI:17409
96%

Chemical Identifiers

1679-53-4
188.27
YJCJVMMDTBEITC-UHFFFAOYSA-N
74300
10-hydroxydecanoic acid
C10H20O3
MFCD00010510
10-hydroxydecanoate, decanoic acid, 10-hydroxy, unii-np03xo416b, 10-hydroxy capric acid, 10-hydroxy-decanoic acid, 1-hydroxydecanoicacid, 10-oh-capric acid, 10-hydroxycapric acid, 10-oh-decanoic acid, 10-hydroxydecanoicacid
CHEBI:17409
OCCCCCCCCCC(O)=O
Safety and Handling

Safety and Handling

EINECSNumber : 216-848-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only