Please login to your online account to display your discounted pricing

Alfa Aesar™ 10-Hydroxybenzo[h]quinoline, 98%

$117.14 - $434.08

Chemical Identifiers

Chemical Name or Material 10-Hydroxybenzo[h]quinoline
CAS 33155-90-7
Molecular Formula C13H9NO
Formula Weight 195.22
MDL Number MFCD00142350
Beilstein 141770
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1966906

  SDS
alfa aesar™
L1966906
5g Each for $117.14

AAL1966914

  SDS
alfa aesar™
L1966914
25g Each for $434.08
Description & Specifications

Specifications

Formula Weight 195.22
MDL Number MFCD00142350
CAS 33155-90-7
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 10-Hydroxybenzo[h]quinoline
Molecular Formula C13H9NO
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Beilstein 141770
Health Hazard 1 Warning
Melting Point 103° to 105°C
Assay 98%

Applications
10-Hydroxybenzo[h]quinolines is used to detect boronic acids in reactions and boron containing compounds on solid support. It is also used as a proton transfer-fluorescence probe to be applied to human serum albumin and beaver apomyoglobin.

Solubility
Soluble in acetone, chloroform and ethyl acetate.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.