missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
10-Hexylphenothiazine, TCI America™
Chemical Identifiers
CAS |
73025-93-1 |
Molecular Formula |
C18H21NS |
Molecular Weight (g/mol) |
283.433 |
InChI Key |
VRJVOTTZESKWPD-UHFFFAOYSA-N |
PubChem CID |
18972631 |
IUPAC Name |
10-hexylphenothiazine |
SMILES |
CCCCCCN1C2=CC=CC=C2SC3=CC=CC=C31 |
View More Specs
Products
2
Specifications
Chemical Identifiers
|
73025-93-1 |
|
283.433 |
|
18972631 |
|
CCCCCCN1C2=CC=CC=C2SC3=CC=CC=C31 |
|
C18H21NS |
|
VRJVOTTZESKWPD-UHFFFAOYSA-N |
|
10-hexylphenothiazine |
Specifications
|
Yellow |
|
73025-93-1 |
|
VRJVOTTZESKWPD-UHFFFAOYSA-N |
|
10-hexylphenothiazine |
|
18972631 |
|
Clear Liquid at 20°C |
|
10-Hexylphenothiazine |
|
5g |
|
C18H21NS |
|
CCCCCCN1C2=CC=CC=C2SC3=CC=CC=C31 |
|
283.433 |
|
283.43 |
|
≥98.0% (GC) |
Safety and Handling