Please login to your online account to display your discounted pricing

Alfa Aesar™ 10-Deacetylbaccatin III, 95%

An intermediate in the synthesis of paclitaxel

Chemical Identifiers

Chemical Name or Material 10-Deacetylbaccatin III
CAS 32981-86-5
Formula Weight 544.59
MDL Number MFCD00132913
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66228LB0

 
alfa aesar™
J66228LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66228MB

 
alfa aesar™
J66228MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 544.59
Melting Point 229° to 236°C
Chemical Name or Material 10-Deacetylbaccatin III
Assay 95%
CAS 32981-86-5
Physical Form Powder
Boiling Point 717°C
MDL Number MFCD00132913
Color White
Quantity 5mg
Solubility Information Soluble in chloroform; Also soluble in methanol with warming; Insoluble in water
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C