Please login to your online account to display your discounted pricing

Alfa Aesar™ 10-Chloro-3-decyne, 98%

$73.89 - $295.54

Chemical Identifiers

Chemical Name or Material 10-Chloro-3-decyne
CAS 18295-64-2
Molecular Formula ClCH2(CH2)5C≡CCH2CH3
Formula Weight 172.7
MDL Number MFCD01321154
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2155606

 
alfa aesar™
B2155606
5g Each for $73.89

AAB2155614

 
alfa aesar™
B2155614
25g Each for $295.54
Description & Specifications

Specifications

Formula Weight 172.7
Chemical Name or Material 10-Chloro-3-decyne
TSCA TSCA
Assay 98%
Molecular Formula ClCH2(CH2)5C≡CCH2CH3
MDL Number MFCD01321154
CAS 18295-64-2
Flash Point >110°C (230°F)
Boiling Point 122°C (20mmHg)
Quantity 5g