missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

10,12-Tricosadiynoic acid, 96%, Thermo Scientific Chemicals

$101.11 - $332.37

Chemical Identifiers

CAS 66990-30-5
Molecular Formula C23H38O2
Molecular Weight (g/mol) 346.555
MDL Number MFCD00041683
InChI Key DIEDVCMBPCRJFQ-UHFFFAOYSA-N
Synonym 10,12-tricosadiynoic acid, 10,12-tricosadiynoicacid, acmc-209nx8, 10,12-tricosadiynoic acid gc
PubChem CID 538457
IUPAC Name tricosa-10,12-diynoic acid
SMILES CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1023103
View Documents Promotion Details
Thermo Scientific Chemicals
L1023103
1 g
Each for $101.11
 
AAL1023106
View Documents Promotion Details
Thermo Scientific Chemicals
L1023106
5 g
Each for $332.37
 
Description

Description

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

66990-30-5
346.555
DIEDVCMBPCRJFQ-UHFFFAOYSA-N
538457
CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
C23H38O2
MFCD00041683
10,12-tricosadiynoic acid, 10,12-tricosadiynoicacid, acmc-209nx8, 10,12-tricosadiynoic acid gc
tricosa-10,12-diynoic acid

Specifications

66990-30-5
C23H38O2
1 g
Light sensitive
Insoluble in water.
CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
346.555
346.56
10,12-Tricosadiynoic acid
55°C to 59°C
MFCD00041683
1880414
10,12-tricosadiynoic acid, 10,12-tricosadiynoicacid, acmc-209nx8, 10,12-tricosadiynoic acid gc
DIEDVCMBPCRJFQ-UHFFFAOYSA-N
tricosa-10,12-diynoic acid
538457
96%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only