missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

10,12-Heptacosadiynoic Acid, TCI America™

$218.48 - $647.80

Chemical Identifiers

CAS 67071-94-7
Molecular Formula C27H46O2
Molecular Weight (g/mol) 402.663
MDL Number MFCD00060065
InChI Key DWPBEQMBEZDLDV-UHFFFAOYSA-N
PubChem CID 20064705
IUPAC Name heptacosa-10,12-diynoic acid
SMILES CCCCCCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H0666500MG View Documents TCI America
H0666500MG
500mg Each for $647.80
 
H0666100MG View Documents TCI America
H0666100MG
100mg Each for $218.48
 
Specifications

Chemical Identifiers

67071-94-7
402.663
DWPBEQMBEZDLDV-UHFFFAOYSA-N
heptacosa-10,12-diynoic acid
C27H46O2
MFCD00060065
20064705
CCCCCCCCCCCCCCC#CC#CCCCCCCCCC(=O)O

Specifications

Undesignated
500mg
C27H46O2
DWPBEQMBEZDLDV-UHFFFAOYSA-N
heptacosa-10,12-diynoic acid
20064705
Crystal-Powder at 20°C
10,12-Heptacosadiynoic Acid
69°C
67071-94-7
MFCD00060065
CCCCCCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
402.663
402.66
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator