missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

10-(1,1′:3′,1′′-Terphenyl-5′-yl)anthracene-9-boronic Acid, TCI America™

(contains varying amounts of Anhydride)

Manufacturer:  TCI America T32801G

 View more versions of this product

Catalog No. T32801G



Specifications

Specifications

10-(1,1′:3′,1′′-Terphenyl-5′-yl)anthracene-9-boronic Acid (contains varying amounts of Anhydride)
1g
C32H23BO2
NQOSAYPSOKFYBW-UHFFFAOYSA-N
[10-(3,5-diphenylphenyl)anthracen-9-yl]boronic acid
127255460
Crystal-Powder at 20°C
Yellow
1415334-59-6
10-(m-Terphenyl-5′C-yl)anthracene-9-boronic Acid
B(C1=C2C=CC=CC2=C(C3=CC=CC=C13)C4=CC(=CC(=C4)C5=CC=CC=C5)C6=CC=CC=C6)(O)O
450.344
450.34
≥97.0% (HPLC)

Chemical Identifiers

1415334-59-6
450.344
10-(m-Terphenyl-5′C-yl)anthracene-9-boronic Acid
[10-(3,5-diphenylphenyl)anthracen-9-yl]boronic acid
C32H23BO2
NQOSAYPSOKFYBW-UHFFFAOYSA-N
127255460
B(C1=C2C=CC=CC2=C(C3=CC=CC=C13)C4=CC(=CC(=C4)C5=CC=CC=C5)C6=CC=CC=C6)(O)O
Safety and Handling

Safety and Handling

TSCA : No

Documents