Learn More
1-Vinylnaphthalene, 95%, stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5104303
Description
1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.
Solubility
Soluble in ethyl acetate, hexane and methanol.
Notes
Moisture sensitive. Incompatible with oxidizing agents, heat, light, ultraviolet radiation and free radical initiators.
Specifications
1-Vinylnaphthalene | |
826-74-4 | |
135°C to 138°C | |
MFCD00075766 | |
Moisture sensitive | |
Soluble in ethyl acetate,hexane and methanol. | |
C=CC1=C2C=CC=CC2=CC=C1 | |
154.21 | |
CHEBI:51327 | |
95% |
stabilized with 4-tert-butylcatechol | |
1.04 | |
C12H10 | |
1 g | |
1-vinylnaphthalene, vinylnaphthalene, 1-vinyl naphthalene, unii-pf20cj2k0n, pf20cj2k0n, 1-vinylphthalene, 1-vinyl-naphthalene, naphthalene, ethenyl, naphthalen-1-yl ethene, naphthalene, 1-ethenyl | |
IGGDKDTUCAWDAN-UHFFFAOYSA-N | |
1-ethenylnaphthalene | |
70004 | |
154.21 |
Chemical Identifiers
826-74-4 | |
154.21 | |
IGGDKDTUCAWDAN-UHFFFAOYSA-N | |
70004 | |
1-ethenylnaphthalene |
C12H10 | |
MFCD00075766 | |
1-vinylnaphthalene, vinylnaphthalene, 1-vinyl naphthalene, unii-pf20cj2k0n, pf20cj2k0n, 1-vinylphthalene, 1-vinyl-naphthalene, naphthalene, ethenyl, naphthalen-1-yl ethene, naphthalene, 1-ethenyl | |
CHEBI:51327 | |
C=CC1=C2C=CC=CC2=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 212-560-5
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only