missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Undecene, 97%

Manufacturer:  Alfa Aesar™ H5338706

 View more versions of this product

Catalog No. AAH5338706



Description

Description

Applications
1-Undecene was used as a biomarker for identification of Pseudomonas aeruginosa strain. It is also used as pharmaceutical intermediate.

Solubility
Solunle in ether, chloroform and ligroin. Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

1-Undecene
192°C to 193°C
5g
C11H22
MFCD00008956
1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720
CCCCCCCCCC=C
154.297
CHEBI:77444
97%
63°C (145°F)
-49°C
821-95-4
CH3(CH2)8CH=CH2
1740044
DCTOHCCUXLBQMS-UHFFFAOYSA-N
undec-1-ene
13190
154.3

Chemical Identifiers

821-95-4
154.297
DCTOHCCUXLBQMS-UHFFFAOYSA-N
13190
undec-1-ene
C11H22
MFCD00008956
1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720
CHEBI:77444
CCCCCCCCCC=C
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents