Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Undecanol, 98%

$58.95 - $58.95

Chemical Identifiers

Chemical Name or Material 1-Undecanol
CAS 112-42-5
Molecular Formula C11H24O
Linear Formula CH3(CH2)10OH
Formula Weight 172.31
MDL Number MFCD00004751
UN Number UN3082
Beilstein 1698334
Synonym n-Undecyl alcohol; 1-Hydroxyundecane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1781722

  SDS
alfa aesar™
A1781722*
100g Each for $58.95

AAA1781736

  SDS
alfa aesar™
A1781736
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.31
MDL Number MFCD00004751
CAS 112-42-5
Flash Point 113°C (235°F)
RTECS Number YQ3155000
Chemical Name or Material 1-Undecanol
TSCA TSCA
Assay 98%
Molecular Formula C11H24O
Linear Formula CH3(CH2)10OH
UN Number UN3082
Beilstein 1698334
Odor Mild
Synonym n-Undecyl alcohol; 1-Hydroxyundecane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P273-P305+P351+P338
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 13° to 15°C
Boiling Point 243°C
Quantity 100g
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

Applications
Used as a synthetic flavoring for food and in perfumes.

Solubility
Not miscible or difficult to mix with water. Soluble in chloroform and ethyl acetate.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P273-P305+P351+P338
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.