Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Trityl-(R)-3-methylpiperazine, 98%, ee 99%

$78.81 - $227.81

Chemical Identifiers

Chemical Name or Material 1-Trityl-(R)-3-methylpiperazine
CAS 313657-75-9
Molecular Formula C24H26N2
Formula Weight 342.49
MDL Number MFCD03426411
Synonym (R)-2-Methyl-4-(triphenylmethyl)piperazine; (R)-2-Methyl-4-tritylpiperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2007803

 
alfa aesar™
L2007803
1g Each for $227.81

AAL20078MD

 
alfa aesar™
L20078MD
250mg Each for $78.81
Description & Specifications

Specifications

Formula Weight 342.49
Assay 98%
Chemical Name or Material 1-Trityl-(R)-3-methylpiperazine
Molecular Formula C24H26N2
MDL Number MFCD03426411
CAS 313657-75-9
Synonym (R)-2-Methyl-4-(triphenylmethyl)piperazine; (R)-2-Methyl-4-tritylpiperazine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 128° to 129°C
Assay Percent Notes ee 99%
Quantity 1g
Optical Rotation −7.6° (c=1 in MeOH)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.