Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Trimethylsilyl-2-pyrrolidinone, 96%


Quantity 10g

Manufacturer: alfa aesar™  L0263809

 View more versions of this product

Catalog No. AAL0263809

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 157.29
Chemical Name or Material 1-Trimethylsilyl-2-pyrrolidinone
Health Hazard 2 GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C7H15NOSi
Health Hazard 1 Warning
Assay 96%
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Synonym N-Trimethylsilylbutyrolactam; N-Trimethylsilyl-2-pyrrolidone
Quantity 10g
Flash Point 46°C (115°F)
UN Number UN1993
Boiling Point 90°C (20mmHg)
CAS 14468-90-7
Beilstein 114950
MDL Number MFCD00051572

Specifications

Chemical Name or Material 1-Trimethylsilyl-2-pyrrolidinone
CAS 14468-90-7
Molecular Formula C7H15NOSi
Formula Weight 157.29
MDL Number MFCD00051572
UN Number UN1993
Beilstein 114950
Synonym N-Trimethylsilylbutyrolactam; N-Trimethylsilyl-2-pyrrolidone

Applications
1-Trimethylsilyl-2-pyrrolidinone, is used as a fine chemical intermediate.

Solubility
It hydrolyzes with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Accessories

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents