Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Trimethylsilyl-2-pyrrolidinone, 96%

Chemical Identifiers

Chemical Name or Material 1-Trimethylsilyl-2-pyrrolidinone
CAS 14468-90-7
Molecular Formula C7H15NOSi
Formula Weight 157.29
MDL Number MFCD00051572
UN Number UN1993
Beilstein 114950
Synonym N-Trimethylsilylbutyrolactam; N-Trimethylsilyl-2-pyrrolidone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0263809

 
alfa aesar™
L0263809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0263818

 
alfa aesar™
L0263818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 157.29
Chemical Name or Material 1-Trimethylsilyl-2-pyrrolidinone
Molecular Formula C7H15NOSi
Health Hazard 2 GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Assay 96%
Synonym N-Trimethylsilylbutyrolactam; N-Trimethylsilyl-2-pyrrolidone
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Quantity 10g
MDL Number MFCD00051572
CAS 14468-90-7
UN Number UN1993
Beilstein 114950
Flash Point 46°C (115°F)
Boiling Point 90°C (20mmHg)

Applications
1-Trimethylsilyl-2-pyrrolidinone, is used as a fine chemical intermediate.

Solubility
It hydrolyzes with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.