missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Trimethylsilyl)-2-naphthyl Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America T24651G

 View more versions of this product

Catalog No. T24651G



Specifications

Specifications

1-(Trimethylsilyl)-2-naphthyl Trifluoromethanesulfonate
1g
C14H15F3O3SSi
1-(Trimethylsilyl)-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 1-(Trimethylsilyl)-2-naphthyl Ester
C[Si](C)(C)C1=C(C=CC2=CC=CC=C21)OS(=O)(=O)C(F)(F)F
348.411
348.41
≥96.0% (GC)
Yellow
252054-88-9
3265
DHFMFICBVHCWGQ-UHFFFAOYSA-N
(1-trimethylsilylnaphthalen-2-yl) trifluoromethanesulfonate
11078565
Clear Liquid at 20°C

Chemical Identifiers

252054-88-9
348.411
1-(Trimethylsilyl)-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 1-(Trimethylsilyl)-2-naphthyl Ester
(1-trimethylsilylnaphthalen-2-yl) trifluoromethanesulfonate
C14H15F3O3SSi
DHFMFICBVHCWGQ-UHFFFAOYSA-N
11078565
C[Si](C)(C)C1=C(C=CC2=CC=CC=C21)OS(=O)(=O)C(F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents