Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Trimethylsilyl-1-propyne, 98%

$60.27 - $60.27

Chemical Identifiers

Chemical Name or Material 1-Trimethylsilyl-1-propyne
CAS 6224-91-5
Molecular Formula CH3C≡CSi(CH3)3
Formula Weight 112.25
MDL Number MFCD00009271
UN Number UN1993
Beilstein 1071311
Synonym Methyl trimethylsilyl acetylene; Propargyltrimethylsilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2487503

  SDS
alfa aesar™
B2487503
1g Each for $60.27

AAB2487514

  SDS
alfa aesar™
B2487514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2487506

  SDS
alfa aesar™
B2487506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.25
Molecular Formula CH3C≡CSi(CH3)3
MDL Number MFCD00009271
CAS 6224-91-5
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Synonym Methyl trimethylsilyl acetylene; Propargyltrimethylsilane
UN Number UN1993
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P243-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Take precautionary measures against static discharge.
Store in a well-ventilated place.
Melting Point −69°C
Quantity 1g
Flash Point −3°C (26°F)
Chemical Name or Material 1-Trimethylsilyl-1-propyne
Boiling Point 99° to 100°C
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Assay 98%
Beilstein 1071311

Applications
1-Trimethylsilyl-1-propyne is employed in the preparation of alkynyl silane derivatives. It is also used to prepare highly substituted indenes using palladium-catalyzed carboannulation. Further, it plays an important role in the preparation of indenones through a rhodium-catalyzed reaction with 2-bromophenylboronic acids.

Solubility
Miscible with toluene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.

Safety and Handling

Danger

GHS P Statement
P210-P243-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Take precautionary measures against static discharge.
Store in a well-ventilated place.

GHS H Statement
H225
Highly flammable liquid and vapour.