missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Trimethylsilyl-1-pentyne, 98%

Manufacturer:  Alfa Aesar™ H5342314

 View more versions of this product

Catalog No. AAH5342314



Specifications

Specifications

1-Trimethylsilyl-1-pentyne
C8H16Si
MFCD00077890
1744218
CABCDUQQPBAHEE-UHFFFAOYSA-N
trimethyl(pent-1-ynyl)silane
2760860
98%
25g
18270-17-2
CH3CH2CH2C≡CSi(CH3)3
UN1993
1-trimethylsilyl-1-pentyne, 1-trimethylsilyl-1-pentyne, trimethyl pent-1-yn-1-yl silane, trimethyl pent-1-ynyl silane, 1-pentyn-1-yltrimethylsilane, 1-tms-1-pentyne, acmc-209ejs, 1-trimethylsilylpent-1-yne, 1-trimethylsi;lyl-1-pentyne, 1-trimethylsilyl-1-pentyne
CCCC#C[Si](C)(C)C
140.301
140.3
135°C
23°C (73°F)

Chemical Identifiers

18270-17-2
140.301
CABCDUQQPBAHEE-UHFFFAOYSA-N
2760860
CCCC#C[Si](C)(C)C
C8H16Si
MFCD00077890
1-trimethylsilyl-1-pentyne, 1-trimethylsilyl-1-pentyne, trimethyl pent-1-yn-1-yl silane, trimethyl pent-1-ynyl silane, 1-pentyn-1-yltrimethylsilane, 1-tms-1-pentyne, acmc-209ejs, 1-trimethylsilylpent-1-yne, 1-trimethylsi;lyl-1-pentyne, 1-trimethylsilyl-1-pentyne
trimethyl(pent-1-ynyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

SDS Documents