missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Trimethylsilyl-1,4-pentadiyne, 98%

Manufacturer:  Alfa Aesar™ H5343614

 View more versions of this product

Catalog No. AAH5343614



Description

Description

Notes
Incompatible with strong oxidizing agents and ammonia.
Specifications

Specifications

1-Trimethylsilyl-1,4-pentadiyne
C8H12Si
MFCD00066350
1,4-pentadiynyltrimethylsilane
C[Si](C)(C)C#CCC#C
136.269
136.27
42°C (25mmHg)
8°C (46°F)
71789-10-1
HC≡CCH2C≡CSi(CH3)3
UN1993
JKYMNPFNLMDSFG-UHFFFAOYSA-N
trimethyl(penta-1,4-diynyl)silane
144510
98%
25g

Chemical Identifiers

71789-10-1
136.269
JKYMNPFNLMDSFG-UHFFFAOYSA-N
144510
C[Si](C)(C)C#CCC#C
C8H12Si
MFCD00066350
1,4-pentadiynyltrimethylsilane
trimethyl(penta-1,4-diynyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents