missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Triisopropylsilyl)pyrrole, 95%

$48.64 - $523.35

Chemical Identifiers

CAS 87630-35-1
Molecular Formula C13H25NSi
Formula Weight 223.44
MDL Number MFCD00054932
Beilstein 3540663
Synonym TIPS-pyrrole
Chemical Name or Material 1-(Triisopropylsilyl)pyrrole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1315203 SDS Alfa Aesar™
L1315203
1g Each for $48.64
AAL1315206 SDS Alfa Aesar™
L1315206
5g Each for $144.07
AAL1315214 SDS Alfa Aesar™
L1315214
25g Each for $523.35
Specifications

Description

Applications
1-(Triisopropylsilyl)pyrrole may be employed as reagent in perfluoroalkylation and Vilsmeier formylation reactions. It may be used in the preparation of ethyl 2-(2,4-dinitrophenylhydrazono]-3-[ 1-(triisopropylsily1)-pyrrol-2-yflpropanoate; heterocyclic base, 3-nitropyrrole and 3-nitropyrrole, required for the synthesis of 1 -(2′-deoxy-β-D-ribofuranosyl)-3-nitropyrrole. It participates in various electrophilic substitution reactions specifically at β-position, via reaction with various electrophilic reagents (Br+, I+,NO2+,etc).

Solubility
Not miscible in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

87630-35-1
223.44
3540663
1-(Triisopropylsilyl)pyrrole
C13H25NSi
MFCD00054932
TIPS-pyrrole

Specifications

87630-35-1
223.44
3540663
78°C (0.4mmHg)
1g
95%
C13H25NSi
MFCD00054932
TIPS-pyrrole
106°C (222°F)
1-(Triisopropylsilyl)pyrrole

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning