missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Triisopropylsilyl)pyrrole, 95%

Manufacturer:  Alfa Aesar™ L1315203

 View more versions of this product

Catalog No. AAL1315203



Description

Description

Applications
1-(Triisopropylsilyl)pyrrole may be employed as reagent in perfluoroalkylation and Vilsmeier formylation reactions. It may be used in the preparation of ethyl 2-(2,4-dinitrophenylhydrazono]-3-[ 1-(triisopropylsily1)-pyrrol-2-yflpropanoate; heterocyclic base, 3-nitropyrrole and 3-nitropyrrole, required for the synthesis of 1 -(2′-deoxy-β-D-ribofuranosyl)-3-nitropyrrole. It participates in various electrophilic substitution reactions specifically at β-position, via reaction with various electrophilic reagents (Br+, I+,NO2+,etc).

Solubility
Not miscible in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

1-(Triisopropylsilyl)pyrrole
78°C (0.4mmHg)
87630-35-1
MFCD00054932
1-triisopropylsilyl pyrrole, 1-triisopropylsilyl-1h-pyrrole, n-triisopropylsilylpyrrole, 1h-pyrrole, 1-tris 1-methylethyl silyl, tri propan-2-yl-pyrrol-1-ylsilane, 1-tris propan-2-yl silyl-1h-pyrrole, pubchem9194, 1-triisopropylsilylpyrrole, 1-triisoproylsilyl-pyrrole, acmc-1bkd7
CC(C)[Si](C(C)C)(C(C)C)N1C=CC=C1
223.435
223.44
106°C (222°F)
1g
C13H25NSi
3540663
FBQURXLBJJNDBX-UHFFFAOYSA-N
tri(propan-2-yl)-pyrrol-1-ylsilane
145136
95%

Chemical Identifiers

87630-35-1
223.435
FBQURXLBJJNDBX-UHFFFAOYSA-N
145136
CC(C)[Si](C(C)C)(C(C)C)N1C=CC=C1
C13H25NSi
MFCD00054932
1-triisopropylsilyl pyrrole, 1-triisopropylsilyl-1h-pyrrole, n-triisopropylsilylpyrrole, 1h-pyrrole, 1-tris 1-methylethyl silyl, tri propan-2-yl-pyrrol-1-ylsilane, 1-tris propan-2-yl silyl-1h-pyrrole, pubchem9194, 1-triisopropylsilylpyrrole, 1-triisoproylsilyl-pyrrole, acmc-1bkd7
tri(propan-2-yl)-pyrrol-1-ylsilane
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning

Documents