missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one, TCI America™

Manufacturer:  TCI America T30391G

 View more versions of this product

Catalog No. T30391G



Specifications

Specifications

1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one
170°C
181934-30-5
MFCD18632570
NTHGHMCOPNSZIR-UHFFFAOYSA-N
1-[2-tri(propan-2-yl)silylethynyl]-1$l^{3},2-benziodoxol-3-one
10693803
Crystal-Powder at 20°C
White
1g
C18H25IO2Si
TIPS-EBX
CC(C)[Si](C#CI1C2=CC=CC=C2C(=O)O1)(C(C)C)C(C)C
428.385
428.39
≥98.0% (HPLC)

Chemical Identifiers

181934-30-5
428.385
NTHGHMCOPNSZIR-UHFFFAOYSA-N
10693803
CC(C)[Si](C#CI1C2=CC=CC=C2C(=O)O1)(C(C)C)C(C)C
C18H25IO2Si
MFCD18632570
TIPS-EBX
1-[2-tri(propan-2-yl)silylethynyl]-1$l^{3},2-benziodoxol-3-one
Safety and Handling

Safety and Handling

TSCA : No

Documents