missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole, TCI America™

Manufacturer:  TCI America T26245G

 View more versions of this product

Catalog No. T26245G



Specifications

Specifications

1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole
78°C
887144-97-0
MFCD10567056
HVAPLSNCVYXFDQ-UHFFFAOYSA-N
3,3-dimethyl-1-(trifluoromethyl)-1$l^{3},2-benziodoxole
16043572
Crystal-Powder at 20°C
White-Yellow
5g
C10H10F3IO
3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 1-trifluoromethyl-3,3-dimethyl-1,2-benziodoxole, togni's reagent, togni reagent, unii-3ok0e02mt6, 1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl, 1,3-dihydro-3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1,3-dihydro-1lambda3,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1? 3 ,2-benziodaoxole, togni's reagents dimethyl compound mi
CC1(C2=CC=CC=C2I(O1)C(F)(F)F)C
330.089
330.09
≥97.0% (HPLC)

Chemical Identifiers

887144-97-0
330.089
HVAPLSNCVYXFDQ-UHFFFAOYSA-N
16043572
CC1(C2=CC=CC=C2I(O1)C(F)(F)F)C
C10H10F3IO
MFCD10567056
3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 1-trifluoromethyl-3,3-dimethyl-1,2-benziodoxole, togni's reagent, togni reagent, unii-3ok0e02mt6, 1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl, 1,3-dihydro-3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1,3-dihydro-1lambda3,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1? 3 ,2-benziodaoxole, togni's reagents dimethyl compound mi
3,3-dimethyl-1-(trifluoromethyl)-1$l^{3},2-benziodoxole
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents