Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Trifluoromethyl)-1-cyclobutanecarboxylic acid, 95%

$94.31 - $94.31

Chemical Identifiers

Chemical Name or Material 1-(Trifluoromethyl)-1-cyclobutanecarboxylic acid
CAS 277756-45-3
Molecular Formula C6H7F3O2
Formula Weight 168.12
MDL Number MFCD03425197
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25850MC

  SDS
alfa aesar™
H25850MC
100mg Each for $94.31
Description & Specifications

Specifications

Formula Weight 168.12
Chemical Name or Material 1-(Trifluoromethyl)-1-cyclobutanecarboxylic acid
Assay 95%
Molecular Formula C6H7F3O2
MDL Number MFCD03425197
CAS 277756-45-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 96°C (205°F)
Boiling Point 76°C (14mmHg)
Melting Point 24° to 26°C
Quantity 100mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.