missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Trifluoroacetyl)piperidine, TCI America™

Manufacturer:  TCI America T208725G

 View more versions of this product

Catalog No. T208725G



Specifications

Specifications

1-(Trifluoroacetyl)piperidine
Colorless
340-07-8
MFCD00466213
C1CCN(CC1)C(=O)C(F)(F)F
181.158
181.16
≥98.0% (GC)
77°C
25g
C7H10F3NO
BCJUMGSHTLYECD-UHFFFAOYSA-N
2,2,2-trifluoro-1-piperidin-1-ylethanone
5181830
Clear Liquid at 20°C

Chemical Identifiers

340-07-8
181.158
BCJUMGSHTLYECD-UHFFFAOYSA-N
2,2,2-trifluoro-1-piperidin-1-ylethanone
C7H10F3NO
MFCD00466213
5181830
C1CCN(CC1)C(=O)C(F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents