Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(trans-2-Phenylethenyl)-4-(phenylethynyl)benzene, 97%

$497.19 - $497.19

Chemical Identifiers

Chemical Name or Material 1-(trans-2-Phenylethenyl)-4-(phenylethynyl)benzene
CAS 21850-30-6
Molecular Formula C22H16
Formula Weight 280.37
MDL Number MFCD12407077
Synonym 4-(trans-beta-Styryl)diphenylacetylene; 4-Ethynyl-trans-stilbene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3063803

  SDS
alfa aesar™
H3063803
1g Each for $497.19
Description & Specifications

Specifications

Formula Weight 280.37
Chemical Name or Material 1-(trans-2-Phenylethenyl)-4-(phenylethynyl)benzene
Assay 97%
Molecular Formula C22H16
MDL Number MFCD12407077
CAS 21850-30-6
Synonym 4-(trans-beta-Styryl)diphenylacetylene; 4-Ethynyl-trans-stilbene
Melting Point 212° to 214°C
Quantity 1g