Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene, 97%

$497.19 - $497.19

Chemical Identifiers

Chemical Name or Material 1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene
CAS 95166-77-1
Molecular Formula C22H20
Formula Weight 284.4
MDL Number MFCD12407080
Synonym 4-Phenethyl-trans-stilbene; 4-(trans-beta-Styryl)bibenzyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3107703

 
alfa aesar™
H3107703
1g Each for $497.19
Description & Specifications

Specifications

Formula Weight 284.4
Chemical Name or Material 1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene
Assay 97%
Molecular Formula C22H20
MDL Number MFCD12407080
CAS 95166-77-1
Synonym 4-Phenethyl-trans-stilbene; 4-(trans-beta-Styryl)bibenzyl
Melting Point 155° to 157°C
Quantity 1g