missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Thio-beta-D-glucose tetraacetate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L11404MD
Description
1-Thio-beta-D-glucose tetraacetate is used as an inhibitor of the Maillard reaction between glucose and glycine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Thio-beta-D-glucose tetraacetate is used as an inhibitor of the Maillard reaction between glucose and glycine.
Solubility
Soluble in water and chloroform (50 mg/ml).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications
1-Thio-beta-D-glucose tetraacetate | |
115°C to 117°C | |
MFCD00063271 | |
1440689 | |
Soluble in water and chloroform (50mg/ml). | |
+5.5° ± 1° (c=2, Chloroform) | |
[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-sulfanyloxan-2-yl]methyl acetate | |
88293 | |
99% |
19879-84-6 | |
C14H20O9S | |
250 mg | |
1-thio-beta-d-glucose tetraacetate, 2r,3r,4s,5r,6s-2-acetoxymethyl-6-mercaptotetrahydro-2h-pyran-3,4,5-triyl triacetate, 1-thio-ss-d-glucose tetraacetate, 1-thio-beta-d-glucose 2,3,4,6-tetraacetate, 1-thio-b-d-glucose tetraacetate, 1-thio-beta-d-glucose 2,3,4,6-tetraacetate, a-d-glucose tetraacetate, 1-thio-, 2,3,4,6-tetra-o-acetyl-1-thio-, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-sulfanyloxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-1-thio-, a-d-glucopyranose | |
SFOZKJGZNOBSHF-UHFFFAOYNA-N | |
CC(=O)OCC1OC(S)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O | |
364.37 | |
364.37 |
Chemical Identifiers
19879-84-6 | |
364.37 | |
SFOZKJGZNOBSHF-UHFFFAOYNA-N | |
88293 | |
CC(=O)OCC1OC(S)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O |
C14H20O9S | |
MFCD00063271 | |
1-thio-beta-d-glucose tetraacetate, 2r,3r,4s,5r,6s-2-acetoxymethyl-6-mercaptotetrahydro-2h-pyran-3,4,5-triyl triacetate, 1-thio-ss-d-glucose tetraacetate, 1-thio-beta-d-glucose 2,3,4,6-tetraacetate, 1-thio-b-d-glucose tetraacetate, 1-thio-beta-d-glucose 2,3,4,6-tetraacetate, a-d-glucose tetraacetate, 1-thio-, 2,3,4,6-tetra-o-acetyl-1-thio-, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-sulfanyloxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-1-thio-, a-d-glucopyranose | |
[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-sulfanyloxan-2-yl]methyl acetate |
Safety and Handling
EINECSNumber : 243-392-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only