missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, TCI America™

Manufacturer:  TCI America T32151G

 View more versions of this product

Catalog No. T32151G



Specifications

Specifications

1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
76°C
903550-26-5
MFCD09037501
ZZRFDLHBMBHJTI-UHFFFAOYSA-N
1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
11587208
Crystal-Powder at 20°C
White
1g
C14H23BN2O3
1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-2-tetrahydropyranyl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydropyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-y-1h-pyrazole, 1-tetrahydropyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole
B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2C3CCCCO3
278.159
278.16
≥98.0% (GC,T)

Chemical Identifiers

903550-26-5
278.159
ZZRFDLHBMBHJTI-UHFFFAOYSA-N
11587208
B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2C3CCCCO3
C14H23BN2O3
MFCD09037501
1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-2-tetrahydropyranyl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydropyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-y-1h-pyrazole, 1-tetrahydropyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole
1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents