missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(Tetrahydro-2-furoyl)piperazine 97.0+%, TCI America™
Supplier: TCI America T261725G
Specifications
1-(Tetrahydro-2-furoyl)piperazine | |
125°C | |
Clear Liquid at 20°C | |
MFCD00190247 | |
UKESBLFBQANJHH-UHFFFAOYSA-N | |
oxolan-2-yl(piperazin-1-yl)methanone | |
2734641 | |
≥97.0% (GC) |
63074-07-7 | |
25 g | |
C9H16N2O2 | |
1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone | |
C1CC(OC1)C(=O)N2CCNCC2 | |
184.239 | |
184.24 |
Chemical Identifiers
63074-07-7 | |
184.239 | |
UKESBLFBQANJHH-UHFFFAOYSA-N | |
2734641 | |
C1CC(OC1)C(=O)N2CCNCC2 |
C9H16N2O2 | |
MFCD00190247 | |
1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone | |
oxolan-2-yl(piperazin-1-yl)methanone |
Safety and Handling
TSCA : No