missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(Tetrahydro-2-furoyl)piperazine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 367160010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-(Tetrahydro-2-furoyl)piperazine | |
63074-07-7 | |
100.0 | |
140°C (2.0 mbar) | |
97.5% min. (GC) | |
C9H16N2O2 | |
MFCD00190247 | |
1.17 | |
Solubility in water: miscible | |
C1CC(OC1)C(=O)N2CCNCC2 | |
184.24 | |
184.24 |
98% | |
97.5 | |
1.1700g/mL | |
Authentic | |
Glass bottle | |
1.5195 to 1.5215 | |
1g | |
1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone | |
UKESBLFBQANJHH-UHFFFAOYSA-N | |
oxolan-2-yl(piperazin-1-yl)methanone | |
2734641 | |
98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO â Research Use Only