missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Tetradecylphosphonic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1751103

 View more versions of this product

Catalog No. AAA1751103


Description

Description

1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.

Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide. Partially soluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
Specifications

Specifications

1-Tetradecylphosphonic acid
C14H31O3P
1 g
Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide. Partially soluble in water.
CCCCCCCCCCCCCCP(=O)(O)O
278.373
278.37
4671-75-4
MFCD00015834
phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate
BVQJQTMSTANITJ-UHFFFAOYSA-N
tetradecylphosphonic acid
78401
98%

Chemical Identifiers

4671-75-4
278.373
BVQJQTMSTANITJ-UHFFFAOYSA-N
78401
CCCCCCCCCCCCCCP(=O)(O)O
C14H31O3P
MFCD00015834
phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate
tetradecylphosphonic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 225-118-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only