Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Tetradecylboronic acid

$52.59 - $662.97

Chemical Identifiers

Chemical Name or Material 1-Tetradecylboronic acid
CAS 100888-40-2
Molecular Formula CH3(CH2)13B(OH)2
Formula Weight 242.21
MDL Number MFCD00093676
Beilstein 1757986
Synonym Myristylboronic acid; n-Tetradecaneboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1996603

  SDS
alfa aesar™
L1996603
1g Each for $52.59

AAL1996606

  SDS
alfa aesar™
L1996606
5g Each for $170.27

AAL1996614

  SDS
alfa aesar™
L1996614
25g Each for $662.97
Description & Specifications

Specifications

Formula Weight 242.21
Chemical Name or Material 1-Tetradecylboronic acid
Molecular Formula CH3(CH2)13B(OH)2
Health Hazard 1 Warning
Synonym Myristylboronic acid; n-Tetradecaneboronic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Melting Point 72° to 76°C
Beilstein 1757986
CAS 100888-40-2
MDL Number MFCD00093676
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.