Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Tetradecyl isothiocyanate, 95%


Quantity 5g

Manufacturer: alfa aesar™  L10547-06

 View more versions of this product

Catalog No. AAL1054706

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Boiling Point 164° to 165°C (3mmHg)
Odor Irritating
Quantity 5g
Chemical Name or Material 1-Tetradecyl isothiocyanate
CAS 3224-48-4
Assay 95%
Molecular Formula CH3(CH2)13NCS
Formula Weight 255.47g/mol
MDL Number MFCD00041122
UN Number UN1760
Beilstein 1781073
Synonym Myristyl isothiocyanate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.

Specifications

Chemical Name or Material 1-Tetradecyl isothiocyanate
CAS 3224-48-4
Molecular Formula CH3(CH2)13NCS
Formula Weight 255.47g/mol
MDL Number MFCD00041122
UN Number UN1760
Beilstein 1781073
Synonym Myristyl isothiocyanate
Accessories

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

2° to 8°C