missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Tetradecene, 94%, Alfa Aesar™

Manufacturer:  Alfa Aesar A13974AP

 View more versions of this product

Catalog No. AAA13974AP



Description

Description

Applications
1-Tetradecene used in the production of linear plasticizers, oxo-alcohols, motor fuels, lubricants, automotive additives, biodegradable surfactants, paper size, and in a wide range of specialty applications in the production of mercaptans, flavors and fragrances, alkyl metals, halides, alkyl silanes.. It is also used as a co monomers in polymer such as low density polyethylenes for the properties of lighter, thinner, better flexibility and more tearing resistance.

Solubility
Soluble in hydrocarbon solvents; insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

1-Tetradecene
500mL
94%
CH3(CH2)11CH=CH2
1701156
HFDVRLIODXPAHB-UHFFFAOYSA-N
tetradec-1-ene
14260
196.38
0.775
1120-36-1
C14H28
MFCD00008981
1-tetradecene, n-tetradec-1-ene, tetradecene, 1-tetradecylene, alpha-tetradecene, unii-fw23481s7s, ccris 3785, alkenes, c10-16 .alpha., a-tetradecene, tetradecene-1
CCCCCCCCCCCCC=C
196.378
CHEBI:77505
94%

Chemical Identifiers

1120-36-1
196.378
HFDVRLIODXPAHB-UHFFFAOYSA-N
14260
tetradec-1-ene
C14H28
MFCD00008981
1-tetradecene, n-tetradec-1-ene, tetradecene, 1-tetradecylene, alpha-tetradecene, unii-fw23481s7s, ccris 3785, alkenes, c10-16 .alpha., a-tetradecene, tetradecene-1
CHEBI:77505
CCCCCCCCCCCCC=C
Safety and Handling

Safety and Handling

GHS H Statement
H304
May be fatal if swallowed and enters airways.

GHS P Statement
P301+P330+P331-P315-P331
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.
Do NOT induce vomiting.

Danger

TSCA : TSCA

SDS Documents