Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Tetradecene, 94%

$33.24 - $146.08

Chemical Identifiers

Chemical Name or Material 1-Tetradecene
CAS 1120-36-1
Molecular Formula C14H28
Linear Formula CH3(CH2)11CH=CH2
Formula Weight 196.38
MDL Number MFCD00008981
Beilstein 1701156
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA13974AP

 
alfa aesar
A13974AP
500mL Each for $38.18

AAA139740F

 
alfa aesar™
A139740F
2500mL Each for $146.08

AAA13974AE

 
alfa aesar™
A13974AE
100mL Each for $33.24
Description & Specifications

Specifications

Formula Weight 196.38
Quantity 500mL
TSCA TSCA
Assay 94%
Linear Formula CH3(CH2)11CH=CH2
MDL Number MFCD00008981
Beilstein 1701156
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H304
May be fatal if swallowed and enters airways.
Health Hazard 3 GHS P Statement
P301+P330+P331-P315-P331
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.
Do NOT induce vomiting.
Chemical Name or Material 1-Tetradecene
Assay Percent Range 94%
Molecular Formula CH3(CH2)11CH=CH2
CAS 1120-36-1
Density 0.775

Applications
1-Tetradecene used in the production of linear plasticizers, oxo-alcohols, motor fuels, lubricants, automotive additives, biodegradable surfactants, paper size, and in a wide range of specialty applications in the production of mercaptans, flavors and fragrances, alkyl metals, halides, alkyl silanes.. It is also used as a co monomers in polymer such as low density polyethylenes for the properties of lighter, thinner, better flexibility and more tearing resistance.

Solubility
Soluble in hydrocarbon solvents; insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P301+P330+P331-P315-P331
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.
Do NOT induce vomiting.

GHS H Statement
H304
May be fatal if swallowed and enters airways.