Please login to your online account to display your discounted pricing

1-(tert-Butyldimethylsilyloxy)-2-propanone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(tert-Butyldimethylsilyloxy)-2-propanone
CAS 74685-00-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H20O2Si
Formula Weight 188.34
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC431230250

 
thermo fisher scientific
AC431230250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 188.34
Quantity 25mL
Color Clear colorless
Density 0.976g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
CAS 74685-00-0
CAS Min % 97.5
Chemical Name or Material 1-(tert-Butyldimethylsilyloxy)-2-propanone
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C9H20O2Si
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.